Learn More
Cayman Chemical Uracil-13C4,15N2, Purity: ≥98%, CAS: 202407-06-5, 5 mg
SDP
Catalog No. 502942840
Encompass_Preferred
An internal standard for the quantification of uracil by GC- or LC-MS
Catalog No. 50-294-2840 Supplier Cayman Chemical Supplier No. 411255 MG
An internal standard for the quantification of uracil by GC- or LC-MS