Learn More
Apexbio Technology LLC Uracil 66-22-8 1g
SDP
Catalog No. 502240711
Encompass_Preferred
Uracil (CAS 66-22-8) is a pyrimidine nucleobase that serves as a fundamental component of ribonucleic acid (RNA) where it pairs with adenine during transcription In biological systems uracil is involved in the regulation of genetic information flow and RNA metabolism It functions as a precursor or substrate in nucleotide biosynthesis and catabolism pathways In biomedical research uracil is widely utilized to investigate RNA synthesis nucleotide analog interactions and cellular proliferation Additionally it is employed in drug delivery system development and explored as a building block in pharmaceutical synthesis and nucleoside analog design
This item is not returnable. View return policy
Catalog No. 50-224-0711 Supplier Apexbio Technology LLC Supplier No. B18631000
Uracil (CAS 66-22-8) is a pyrimidine nucleobase that serves as a fundamental component of ribonucleic acid (RNA) where it pairs with adenine during transcription In biological systems uracil is involved in the regulation of genetic information flow and RNA metabolism It functions as a precursor or substrate in nucleotide biosynthesis and catabolism pathways In biomedical research uracil is widely utilized to investigate RNA synthesis nucleotide analog interactions and cellular proliferation Additionally it is employed in drug delivery system development and explored as a building block in pharmaceutical synthesis and nucleoside analog design