Learn More
Medchemexpress LLC Uracil | 202407-06-5 | MDL-MFCD01073456 | 98.0% | 118.04 g/mol | 13C4H4 15N2O2 | 10 MG
SDP
Catalog No. 5000297608
Encompass_Preferred
Uracil-13C4,15N2 is an isotopically labeled uracil provided as a solid research standard for analytical and tracer applications. Four carbon atoms are 13C and two nitrogens are 15N; the material is supplied at high purity and intended for use in mass spectrometry, metabolic labeling, and tracer studies.
  • Isotopically labeled with 13C4 and 15N2.
  • High purity: 98.0%.
  • Suitable for mass spectrometry and tracer studies.
  • Available in small research quantities (10 MG).
  • Supplied as a solid for convenient handling and storage.
Catalog No. 50-002-97608 Supplier Medchemexpress LLC Supplier No. HYI0960S310MG
Uracil-13C4,15N2 is an isotopically labeled uracil provided as a solid research standard for analytical and tracer applications. Four carbon atoms are 13C and two nitrogens are 15N; the material is supplied at high purity and intended for use in mass spectrometry, metabolic labeling, and tracer studies.
  • Isotopically labeled with 13C4 and 15N2.
  • High purity: 98.0%.
  • Suitable for mass spectrometry and tracer studies.
  • Available in small research quantities (10 MG).
  • Supplied as a solid for convenient handling and storage.