Learn More
American Radiolabeled Chemicals Inc URACIL 6-3H 1LC
SDP
Catalog No. 50901208
Encompass_Preferred
Uracil-6-3H 1mCi in sterile water, 1 mCi/ml, S.A. 20-30 Ci/mmol, M.W. 112.1
Catalog No. 50-901-208 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 17821MC
Uracil-6-3H 1mCi in sterile water, 1 mCi/ml, S.A. 20-30 Ci/mmol, M.W. 112.1