Learn More
New England Biolabs, Inc. Uracil Glycosylase Inhibitor (UGI) – 200 units
SDP
Catalog No. 50995218
Encompass
The Uracil Glycosylase Inhibitor (UGI) of Bacillus subtilis bacteriophage PBS1 is a small protein (9.5 kDa) which inhibits E. coli uracil-DNA glycosylase (UDG) as well as UDG from other species. Inhibition of UDG occurs by reversible protein binding with a 1:1 UGD:UGI stoichiometry. UGI is capable of dissociating UDG-DNA complexes.
Catalog No. 50-995-218 Supplier New England Biolabs, Inc. Supplier No. M0281S
The Uracil Glycosylase Inhibitor (UGI) of Bacillus subtilis bacteriophage PBS1 is a small protein (9.5 kDa) which inhibits E. coli uracil-DNA glycosylase (UDG) as well as UDG from other species. Inhibition of UDG occurs by reversible protein binding with a 1:1 UGD:UGI stoichiometry. UGI is capable of dissociating UDG-DNA complexes.