Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uradil impurity 1 | 31737-09-4 | 99.8% | 160.56 | 250 MG
SDP

Catalog No. 5000402329
Encompass_Preferred

Uradil impurity 1 is an impurity of Uradil. It is for research use only and not sold to patients.

  • High purity (99.81%)
  • Known molecular weight (160.56) and molecular formula (C5H5ClN2O2)
  • Available as a solid
  • Specific storage conditions for powder and in solvent
  • Soluble in DMSO

Catalog No. 50-004-02329 Supplier Medchemexpress LLC Supplier No. 250MGHY20821
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uradil impurity 1 is an impurity of Uradil. It is for research use only and not sold to patients.

  • High purity (99.81%)
  • Known molecular weight (160.56) and molecular formula (C5H5ClN2O2)
  • Available as a solid
  • Specific storage conditions for powder and in solvent
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.