Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uranyl Nitrate, Hexahydrate, Crystal, Reagent, ACS, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607648 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-607-648 125 g
18-607-647 25 g
18-607-649 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-607-648 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. U1010125GM
Only null left
Add to Cart
Add to Cart
Request a Quote

U1010, 13520-83-7, UO2(NO3)2·6H2O

Spectrum™ Chemical Uranyl Nitrate, Hexahydrate, Crystal, Reagent, ACS is a yellow green crystal and is water soluble as well as being soluble in acetone, ethanol and ether. It is mainly used as a negative stain for viruses in electron microscopy. It also stabilizes nucleic acids and cell membranes in tissue samples and is also vital for nuclear reprocessing. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 13520-83-7
Molecular Formula H14N2O14U
Molecular Weight (g/mol) 504.14
InChI Key WRIBVRZWDPGVQH-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.O=[U]=O.O[N+]([O-])=O.O[N+]([O-])=O

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 98 to 102%
Grade ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.