Learn More
Medchemexpress LLC Urapidil hydrochloride | 64887-14-5 | 99.86% | 25 MG
SDP
Catalog No. 5000450531
Encompass_Preferred
Urapidil hydrochloride is an orally active and blood-brain barrier permeable α1-adrenoceptor antagonist and 5-HT1A receptor agonist. It has a pIC50 of 6.13 against α1-adrenoceptor and 4.38 against α2-adrenoceptor, exhibiting an antihypertensive effect.
  • Orally active and blood-brain barrier permeable
  • Acts as an α1-adrenoceptor antagonist and 5-HT1A receptor agonist
  • Exhibits antihypertensive effect
  • Research grade material with 99.86% purity
  • Supplied as a white to off-white solid
Catalog No. 50-004-50531 Supplier Medchemexpress LLC Supplier No. 25MGHYB0354AR
Urapidil hydrochloride is an orally active and blood-brain barrier permeable α1-adrenoceptor antagonist and 5-HT1A receptor agonist. It has a pIC50 of 6.13 against α1-adrenoceptor and 4.38 against α2-adrenoceptor, exhibiting an antihypertensive effect.
  • Orally active and blood-brain barrier permeable
  • Acts as an α1-adrenoceptor antagonist and 5-HT1A receptor agonist
  • Exhibits antihypertensive effect
  • Research grade material with 99.86% purity
  • Supplied as a white to off-white solid