Learn More
Medchemexpress LLC Urapidil hydrochloride | 64887-14-5 | 99.86% | 50 MG
SDP
Catalog No. 5000407275
Encompass_Preferred
Urapidil hydrochloride is an orally active and blood-brain barrier permeable α1-adrenoceptor antagonist and 5-HT1A receptor agonist. It has a pIC50 of 6.13 against α1-adrenoceptor and 4.38 against α2-adrenoceptor. It demonstrates antihypertensive effects.
  • Orally active and blood-brain barrier permeable
  • Functions as an α1-adrenoceptor antagonist and 5-HT1A receptor agonist
  • Demonstrates antihypertensive effects
  • Features pIC50 of 6.13 against α1-adrenoceptor
  • Features pIC50 of 4.38 against α2-adrenoceptor
Catalog No. 50-004-07275 Supplier Medchemexpress LLC Supplier No. HYB0354A50MG
Urapidil hydrochloride is an orally active and blood-brain barrier permeable α1-adrenoceptor antagonist and 5-HT1A receptor agonist. It has a pIC50 of 6.13 against α1-adrenoceptor and 4.38 against α2-adrenoceptor. It demonstrates antihypertensive effects.
  • Orally active and blood-brain barrier permeable
  • Functions as an α1-adrenoceptor antagonist and 5-HT1A receptor agonist
  • Demonstrates antihypertensive effects
  • Features pIC50 of 6.13 against α1-adrenoceptor
  • Features pIC50 of 4.38 against α2-adrenoceptor