Learn More
Medchemexpress LLC Urapidil hydrochloride | 64887-14-5 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0354A10MM/1ML
Encompass_Preferred
Urapidil hydrochloride is an orally active α1-adrenoceptor antagonist and 5-HT1A receptor agonist with blood-brain barrier permeability. It exhibits pIC50 values of 6.13 against α1-adrenoceptor and 4.38 against α2-adrenoceptor, and demonstrates an antihypertensive effect.
  • Orally active α1-adrenoceptor antagonist
  • 5-HT1A receptor agonist
  • Blood-brain barrier permeability
  • Exhibits antihypertensive effect
  • pIC50 of 6.13 against α1-adrenoceptor
  • pIC50 of 4.38 against α2-adrenoceptor
Catalog No. 50-004-07264