Learn More
Medchemexpress LLC Urapidil hydrochloride | 64887-14-5 | 99.9% | 100 MG
SDP
Catalog No. 5000406112
Encompass_Preferred
Urapidil hydrochloride is an orally active and blood-brain barrier permeability α1-adrenoceptor antagonist and 5-HT1A receptor agonist with a pIC50 of 6.13 and 4.38 against α1- and α2-adrenoceptor, respectively. It shows an antihypertensive effect.
  • Orally active
  • Blood-brain barrier permeability α1-adrenoceptor antagonist
  • 5-HT1A receptor agonist
  • Shows antihypertensive effect
Catalog No. 50-004-06112 Supplier Medchemexpress LLC Supplier No. HYB0354A100MG
Urapidil hydrochloride is an orally active and blood-brain barrier permeability α1-adrenoceptor antagonist and 5-HT1A receptor agonist with a pIC50 of 6.13 and 4.38 against α1- and α2-adrenoceptor, respectively. It shows an antihypertensive effect.
  • Orally active
  • Blood-brain barrier permeability α1-adrenoceptor antagonist
  • 5-HT1A receptor agonist
  • Shows antihypertensive effect