Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Urapidil (hydrochloride) | 64887-14-5 | MFCD00078601 | 100.0% | 10 MG
SDP

Catalog No. 5000297112
Encompass_Preferred

Urapidil hydrochloride is an α1-adrenoceptor antagonist and 5-HT1A receptor agonist used in pharmacological research for its antihypertensive effects. It is supplied as a hydrochloride salt and characterized by high purity and a molecular weight of 423.94 g/mol.

  • Orally active α1-adrenoceptor antagonist and 5-HT1A receptor agonist.
  • Used in adrenergic receptor and antihypertensive research applications.
  • High purity (COA reports 99.97% by LCMS).
  • Molecular weight 423.94 g/mol; formula C20H30ClN5O3.
  • CAS number 64887-14-5 for unambiguous identification.

Catalog No. 50-002-97112 Supplier Medchemexpress LLC Supplier No. HYB0354AR10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Urapidil hydrochloride is an α1-adrenoceptor antagonist and 5-HT1A receptor agonist used in pharmacological research for its antihypertensive effects. It is supplied as a hydrochloride salt and characterized by high purity and a molecular weight of 423.94 g/mol.

  • Orally active α1-adrenoceptor antagonist and 5-HT1A receptor agonist.
  • Used in adrenergic receptor and antihypertensive research applications.
  • High purity (COA reports 99.97% by LCMS).
  • Molecular weight 423.94 g/mol; formula C20H30ClN5O3.
  • CAS number 64887-14-5 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.