Learn More
Medchemexpress LLC Urapidil (hydrochloride) | 64887-14-5 | MFCD00078601 | 99.98% | 100 MG
SDP
Catalog No. 5000406113
Encompass_Preferred
Urapidil hydrochloride (Standard) is an analytical standard intended for research and analytical applications. It is an orally active α1-adrenoceptor antagonist and 5-HT1A receptor agonist, showing an antihypertensive effect.
  • Analytical standard grade
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments
  • Applicable in HPLC
  • Applicable in GC
  • Applicable in MS
Catalog No. 50-004-06113 Supplier Medchemexpress LLC Supplier No. HYB0354AR100MG
Urapidil hydrochloride (Standard) is an analytical standard intended for research and analytical applications. It is an orally active α1-adrenoceptor antagonist and 5-HT1A receptor agonist, showing an antihypertensive effect.
  • Analytical standard grade
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments
  • Applicable in HPLC
  • Applicable in GC
  • Applicable in MS