Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC URAT1 inhibitor 1 50mg | 2268720-62-1 | 50 MG
SDP

Catalog No. 5000307798
Encompass_Preferred

URAT1 inhibitor 1 is a small-molecule uric acid transporter 1 (URAT1) inhibitor (IC50 = 32 nM) supplied as an off-white to light yellow solid for research use. It is soluble in DMSO and available in multiple pack sizes; follow recommended storage conditions for stability.

  • Potent uric acid transporter 1 (URAT1) inhibitor, IC50 32 nM.
  • Off-white to light yellow solid.
  • Solubility in DMSO: 125 mg/mL.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple pack sizes, including small mg quantities and DMSO solutions.

Catalog No. 50-003-07798 Supplier Medchemexpress LLC Supplier No. HY11430950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

URAT1 inhibitor 1 is a small-molecule uric acid transporter 1 (URAT1) inhibitor (IC50 = 32 nM) supplied as an off-white to light yellow solid for research use. It is soluble in DMSO and available in multiple pack sizes; follow recommended storage conditions for stability.

  • Potent uric acid transporter 1 (URAT1) inhibitor, IC50 32 nM.
  • Off-white to light yellow solid.
  • Solubility in DMSO: 125 mg/mL.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple pack sizes, including small mg quantities and DMSO solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.