Learn More
Bioworld URATE OXIDASE (RECOMBINANT)
Supplier:  Bioworld 220607433
Encompass_Preferred
Recombinant Human Urate Oxidase (UOX) Source:Escherichia Coli. Background : Description : Urate oxidase catalyzes the enzymatic oxidation of uric acid into allantoin, an inactive and soluble metabolite. Recombinant Human Urate Oxid...
Catalog No. 50-253-844