Learn More
Enzo Life Sciences URB-602 (50mg). CAS: 565460-15-3
SDP
Supplier:  Enzo Life Sciences BMLEI3480050
Encompass_Preferred
Selectively inhibits monoacylglycerol (MAG) lipase (IC50=28µM) via a noncompetitive mechanism. Does not inhibit FAAH, DAG lipase or COX-2. URB-602 selectively raises levels of 2-arachidonoylglycerol with no change in anandamide concentrationsin cells and tissues. In the same way anandamide levels were markely elevated by the FAAH inhibitor URB597 at 1µM. Alternative name: Biphenyl-3-yl carbamic acid cyclohexyl ester. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (30 mg/ml) or ethanol (5 mg/ml warm). Long Term Storage: Ambient.
Catalog No. 50-201-0735