Learn More
Apexbio Technology LLC URB597(Synonyms: URB-597, KDS-4103, FAAH inhibitor URB597, Cyclohexylcarbamic acid 3'-carbamoylbiphenyl-3-yl ester), 10mM (in 1mL DMSO), CAS: 546141-08-6.
SDP
Catalog No. 5000060494
Encompass_Preferred
URB597 (CAS 546141-08-6) also known as KDS-4103 is a potent and selective inhibitor of fatty acid amide hydrolase (FAAH) an enzyme responsible for intracellular hydrolysis of the endocannabinoid anandamide In vitro assays indicate that URB597 inhibits FAAH activity with IC values of 3 nM in human liver microsomes and 52 nM in rat brain membranes Intraperitoneal administration in rats results in FAAH inhibition within brain tissue (ID 0 15 mg/kg) URB597 exhibits minimal interaction with cannabinoid receptors anandamide transporters or other related receptors ion channels enzymes and transporters This selectivity makes URB597 valuable for investigating endocannabinoid signaling pathways in biomedical research
Catalog No. 50-000-60494 Supplier Apexbio Technology LLC Supplier No. A43725.1
URB597 (CAS 546141-08-6) also known as KDS-4103 is a potent and selective inhibitor of fatty acid amide hydrolase (FAAH) an enzyme responsible for intracellular hydrolysis of the endocannabinoid anandamide In vitro assays indicate that URB597 inhibits FAAH activity with IC values of 3 nM in human liver microsomes and 52 nM in rat brain membranes Intraperitoneal administration in rats results in FAAH inhibition within brain tissue (ID 0 15 mg/kg) URB597 exhibits minimal interaction with cannabinoid receptors anandamide transporters or other related receptors ion channels enzymes and transporters This selectivity makes URB597 valuable for investigating endocannabinoid signaling pathways in biomedical research