Learn More
Medchemexpress LLC HY-100792 10mg Medchemexpress, URB602 CAS:565460-15-3 Purity:>98%
SDP
Catalog No. 501931951
Encompass_Preferred
Medchemexpress, HY-100792 10mg URB602 CAS:565460-15-3 URB602 is a selective monoacylglycerol lipase ( MGL ) inhibitor, which inhibits rat brain MGL with IC 50 of 28±4 μM through a noncompetitive mechanism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1951 Supplier Medchemexpress LLC Supplier No. HY10079210MG
Medchemexpress, HY-100792 10mg URB602 CAS:565460-15-3 URB602 is a selective monoacylglycerol lipase ( MGL ) inhibitor, which inhibits rat brain MGL with IC 50 of 28±4 μM through a noncompetitive mechanism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.