Learn More
Medchemexpress LLC Urb602 | 565460-15-3 | 99.7% | 295.38 g·mol⁻1 | C19H21NO2 | 25 MG
SDP
Catalog No. 5000211324
Encompass_Preferred
URB602 is a small-molecule research reagent that selectively inhibits monoacylglycerol lipase (MGL). It inhibits rat brain MGL with an IC50 of 28±4 μM via a noncompetitive mechanism and is supplied as a high-purity analytical standard for biochemical and preclinical studies.
  • Selective monoacylglycerol lipase inhibitor with IC50 28±4 μM.
  • Noncompetitive inhibition mechanism useful for enzymology studies.
  • High purity suitable for analytical and biochemical assays.
  • Available in small milligram quantities and solution formats for reconstitution.
  • Chemical formula C19H21NO2; molecular weight 295.38 g·mol⁻1.
Catalog No. 50-002-11324 Supplier Medchemexpress LLC Supplier No. HY10079225MG
URB602 is a small-molecule research reagent that selectively inhibits monoacylglycerol lipase (MGL). It inhibits rat brain MGL with an IC50 of 28±4 μM via a noncompetitive mechanism and is supplied as a high-purity analytical standard for biochemical and preclinical studies.
  • Selective monoacylglycerol lipase inhibitor with IC50 28±4 μM.
  • Noncompetitive inhibition mechanism useful for enzymology studies.
  • High purity suitable for analytical and biochemical assays.
  • Available in small milligram quantities and solution formats for reconstitution.
  • Chemical formula C19H21NO2; molecular weight 295.38 g·mol⁻1.