Learn More
Abcam URB754, Monoacylglycerol lipase inhibitor, 10MG
Catalog No. 502707125
Encompass_Preferred
MW 266.3 g/mol, Purity >98%. Reported potent and non-competitive monoacylglycerol lipase (MAGL) inhibitor (IC₅₀ = 200 nM). Inhibits FAAH (IC₅₀ = 32 μM) and binds weakly to the rat CB₁ receptor (IC₅₀ = 3.8 μM). Shows vasodilatory activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7125 Supplier Abcam Supplier No. AB12092710MG
MW 266.3 g/mol, Purity >98%. Reported potent and non-competitive monoacylglycerol lipase (MAGL) inhibitor (IC₅₀ = 200 nM). Inhibits FAAH (IC₅₀ = 32 μM) and binds weakly to the rat CB₁ receptor (IC₅₀ = 3.8 μM). Shows vasodilatory activity.
The product is subject to the following: Abcam Restricted Use Statement