Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Urban Water Quality Test Kit
SDPSCI_ED

Catalog No. S66226 Shop All LaMotte Company Products
Change view
Click to view available options
Grade:
3+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S66226 3+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S66226 Supplier LaMotte™ Supplier No. 5918
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Materials for 10 tests for each factor (coliform contains five tests), comprehensive handbook, all necessary equipment

Introduces water chemistry concepts in urban areas.

  • Introduces water chemistry concepts in areas where the lifestyle does not often include involvement in outdoor activities or contact with nature
  • Helps urban students understand that environmental issues are a part of their world, and issues involving water chemistry are all around them
  • Employs methods suitable for testing drinking water, runoff and street puddles
  • Tests for chlorine, copper, iron, hardness, nitrate, pH, phosphates, dissolved oxygen, coliform and temperature using nonhazardous TesTabs™

Specifications

pH Range 4 to 10
Volume (Metric) 40 oz.
For Use With (Application) Introduces water chemistry concepts in urban areas
Grade 3+
Includes Materials for 10 tests for each factor, comprehensive handbook, all necessary equipment
Type Urban Water Quality Test Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.