Learn More
Sigma Aldrich Fine Chemicals Biosciences Urea, 57-13-6, MFCD00008022, 100 g
Catalog No. NC0768148
Encompass
Linear Formula: NH2CONH2, Molecular Weight: 60.06, powder, BioReagent, for molecular biology, suitable for cell culture, Synonym: Carbamide, Carbonyldiamide. Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.
Catalog No. NC0768148 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. U5378100G
Linear Formula: NH2CONH2, Molecular Weight: 60.06, powder, BioReagent, for molecular biology, suitable for cell culture, Synonym: Carbamide, Carbonyldiamide. Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.