Learn More
Medchemexpress LLC HY-Y0271 100mg Medchemexpress, Urea CAS:57-13-6 Purity:>98%
SDP
Catalog No. 501934683
Encompass_Preferred
Medchemexpress, HY-Y0271 100mg Urea CAS:57-13-6 Urea is a powerful protein denaturant via both direct and indirect mechanisms [1] . A potent emollient and keratolytic agent [2] . Used as a diuretic agent. Blood urea nitrogen (BUN) has been utilized to evaluate renal function [3] . Widely used in fertilizers as a source of nitrogen and is an important raw material for the chemical industry. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4683 Supplier Medchemexpress LLC Supplier No. HYY0271100MG
Medchemexpress, HY-Y0271 100mg Urea CAS:57-13-6 Urea is a powerful protein denaturant via both direct and indirect mechanisms [1] . A potent emollient and keratolytic agent [2] . Used as a diuretic agent. Blood urea nitrogen (BUN) has been utilized to evaluate renal function [3] . Widely used in fertilizers as a source of nitrogen and is an important raw material for the chemical industry. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.