Learn More
Medchemexpress LLC Urea-13C,15N2 | 58069-83-3 | 99.9% | 63.03 | 5 MG
SDP
Catalog No. 5000393429
Encompass_Preferred
Urea-13C,15N2 is the 13C and 15N labeled Urea, which functions as a powerful protein denaturant through direct and indirect mechanisms. It also acts as a potent emollient and keratolytic agent, and is utilized as a diuretic. It is widely applied in fertilizers as a nitrogen source and serves as an important raw material for the chemical industry. Blood urea nitrogen (BUN) is used to evaluate renal function.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.
Catalog No. 50-003-93429 Supplier Medchemexpress LLC Supplier No. 5MGHYY0271S2
Urea-13C,15N2 is the 13C and 15N labeled Urea, which functions as a powerful protein denaturant through direct and indirect mechanisms. It also acts as a potent emollient and keratolytic agent, and is utilized as a diuretic. It is widely applied in fertilizers as a nitrogen source and serves as an important raw material for the chemical industry. Blood urea nitrogen (BUN) is used to evaluate renal function.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.