Learn More
Medchemexpress LLC Urea-13C, 15N2 | 58069-83-3 | MFCD00145506 | 100.0% | 63.03 | 13CH4 15N2O | 25mg
SDP
Catalog No. 5000267337
Encompass_Preferred
Urea-13C 15N2 is the 13C and 15N labeled Urea[1] Urea is a powerful protein denaturant via both direct and indirect mechanisms[1] A potent emollient and keratolytic agent[2] Used as a diuretic agent Blood urea nitrogen (BUN) has been utilized to evaluate renal function[3] Widely used in fertilizers as a source of nitrogen and is an important raw material for the chemical industry
Catalog No. 50-002-67337 Supplier Medchemexpress LLC Supplier No. HYY0271S225MG
Urea-13C 15N2 is the 13C and 15N labeled Urea[1] Urea is a powerful protein denaturant via both direct and indirect mechanisms[1] A potent emollient and keratolytic agent[2] Used as a diuretic agent Blood urea nitrogen (BUN) has been utilized to evaluate renal function[3] Widely used in fertilizers as a source of nitrogen and is an important raw material for the chemical industry