Learn More
Medchemexpress LLC Urea-13C,15N2 | 58069-83-3 | MFCD00145506 | 100.0% | 63.03 | 13CH415N2O | 10mg
SDP
Catalog No. 5000267859
Encompass_Preferred
Urea-13C 15N2 is the 13C and 15N labeled Urea[1] Urea is a powerful protein denaturant via both direct and indirect mechanisms[1] A potent emollient and keratolytic agent[2] Used as a diuretic agent Blood urea nitrogen (BUN) has been utilized to evaluate renal function[3] Widely used in fertilizers as a source of nitrogen and is an important raw material for the chemical industry
Catalog No. 50-002-67859 Supplier Medchemexpress LLC Supplier No. HYY0271S210MG
Urea-13C 15N2 is the 13C and 15N labeled Urea[1] Urea is a powerful protein denaturant via both direct and indirect mechanisms[1] A potent emollient and keratolytic agent[2] Used as a diuretic agent Blood urea nitrogen (BUN) has been utilized to evaluate renal function[3] Widely used in fertilizers as a source of nitrogen and is an important raw material for the chemical industry