Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea-13C,15N2 Solution NMR Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111002796 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2796 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2796 Supplier MilliporeSigma Supelco Supplier No. 61138710X7IN
Add to Cart
Add to Cart
Request a Quote

Indirect detection

TRUSTED_SUSTAINABILITY

Specifications

Packaging Each. tube contains 0.700 mL. Each Pk. contains 10 tubes.
Concentration 0.1 M in DMSO- d 6 (99.9 atom% D)
Linear Formula 13 CH415N2 O
Type NMR Reference Standard
Quantity 1 Ea.
Formula Weight 63.03
Grade NMR Reference Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.