Learn More
Medchemexpress LLC Urea-13C | 58069-82-2 | MFCD00064398 | 99.1% | 61.05 | 13CH4N2O | 100 MG
SDP
Catalog No. 5000229863
Encompass_Preferred
Urea-13C is a carbon-13 labeled urea supplied as a high-purity solid for diagnostic breath testing and as an isotopic tracer or internal standard in analytical techniques such as NMR, GC-MS, and LC-MS. It is provided in small, pre-measured pack sizes suitable for laboratory and clinical use.
  • High purity (99.1%) suitable for analytical and diagnostic applications.
  • 13C-labeled at the carbonyl position for use in 13C-urea breath testing.
  • Solid white to off-white powder, easy to weigh and dissolve.
  • Molecular weight 61.05, molecular formula 13CH4N2O.
  • Available in 100 MG packages for diagnostic or research use.
Catalog No. 50-002-29863 Supplier Medchemexpress LLC Supplier No. HY118877100MG
Urea-13C is a carbon-13 labeled urea supplied as a high-purity solid for diagnostic breath testing and as an isotopic tracer or internal standard in analytical techniques such as NMR, GC-MS, and LC-MS. It is provided in small, pre-measured pack sizes suitable for laboratory and clinical use.
  • High purity (99.1%) suitable for analytical and diagnostic applications.
  • 13C-labeled at the carbonyl position for use in 13C-urea breath testing.
  • Solid white to off-white powder, easy to weigh and dissolve.
  • Molecular weight 61.05, molecular formula 13CH4N2O.
  • Available in 100 MG packages for diagnostic or research use.