Learn More
Medchemexpress LLC Urea-13C (carbonyl diamide-13C) | 58069-82-2 | MFCD00064398 | 99.1% | 61.05 g/mol | 13CH4N2O | 50 MG
SDP
Catalog No. 5000228626
Encompass_Preferred
Urea-13C is urea labeled with the stable isotope carbon-13 (13C), provided as a solid reagent for diagnostic and analytical applications. It is used as the substrate in the 13C-urea breath test for detection of Helicobacter pylori and as a tracer or internal standard for quantitative NMR, GC-MS, and LC-MS analyses.
  • 13C enrichment of 99.1%.
  • Suitable as substrate for 13C-urea breath testing to detect Helicobacter pylori.
  • Useful as tracer and internal standard for NMR, GC-MS, and LC-MS quantitative analysis.
  • Solid, white to off-white physical form for easy handling and storage.
  • Available in small pack sizes such as 50 mg for research use.
Catalog No. 50-002-28626 Supplier Medchemexpress LLC Supplier No. HY11887750MG
Urea-13C is urea labeled with the stable isotope carbon-13 (13C), provided as a solid reagent for diagnostic and analytical applications. It is used as the substrate in the 13C-urea breath test for detection of Helicobacter pylori and as a tracer or internal standard for quantitative NMR, GC-MS, and LC-MS analyses.
  • 13C enrichment of 99.1%.
  • Suitable as substrate for 13C-urea breath testing to detect Helicobacter pylori.
  • Useful as tracer and internal standard for NMR, GC-MS, and LC-MS quantitative analysis.
  • Solid, white to off-white physical form for easy handling and storage.
  • Available in small pack sizes such as 50 mg for research use.