Learn More
Urea-15N2 Solution NMR Reference Standard, 0.1M in DMSO-d6 (99.9 atom D), NMR tube size 5 mm, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Packaging | Each tube contains 0.700 mL. Each Pk. contains 10 tubes. |
| Concentration | 0.1 M in DMSO- d 6 (99.9 atom% D) |
| Linear Formula | CH415N2 O |
| Type | NMR Reference Standard |
| Quantity | 1 Ea. |
| Formula Weight | 62.04 |
| Grade | NMR Reference Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.