Learn More
Apexbio Technology LLC Urea,1kg CAS# 57-13-6
SDP
Catalog No. 502240514
Encompass_Preferred
Urea is an organic compound generated by the urea cycle enzymes, which is then eliminated through fluids, especially urine. Blood urea nitrogen has been utilized to evaluate renal function for decades. In addition, urea has been used clinically for a variety of dermatological diseases over a century. Urea is a potent emollient and keratolytic agent, making it an effective topical monotherapy for conditions such as dry and scaly skin. Due to its high solubility in aqueous solution, urea further extends its applications in laboratory, serving as a powerful protein denaturant for assessing protein stability, the effects of mutations on stability, and protein unfolding.  Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0514 Supplier Apexbio Technology LLC Supplier No. B77811000000
Urea is an organic compound generated by the urea cycle enzymes, which is then eliminated through fluids, especially urine. Blood urea nitrogen has been utilized to evaluate renal function for decades. In addition, urea has been used clinically for a variety of dermatological diseases over a century. Urea is a potent emollient and keratolytic agent, making it an effective topical monotherapy for conditions such as dry and scaly skin. Due to its high solubility in aqueous solution, urea further extends its applications in laboratory, serving as a powerful protein denaturant for assessing protein stability, the effects of mutations on stability, and protein unfolding.  Other sizes are also available. Please inqury us for quote.