Learn More
Selleck Chemical LLC Urea S3687-25mg
SDP
Catalog No. 503108537
Encompass_Preferred
Urea (Carbonyldiamide Ureophil) is a highly soluble organic compound formed in the liver from ammonia produced by the deamination of amino acids It is the principal end product of protein catabolism and constitutes about one half of the total urinary solids
Catalog No. 50-310-8537 Supplier Selleck Chemical LLC Supplier No. S368725MG
Urea (Carbonyldiamide Ureophil) is a highly soluble organic compound formed in the liver from ammonia produced by the deamination of amino acids It is the principal end product of protein catabolism and constitutes about one half of the total urinary solids