Learn More
Research Products International Corp Urea, UltraPure (USP Grade), 500 Grams
SDP
Catalog No. 501368682
Encompass_Preferred
Urea (CO(NH₂)₂) is a water-soluble compound (60.06 g/mol) used in life science labs as a chaotropic agent to denature proteins and nucleic acids. It’s commonly applied in SDS-PAGE, protein solubilization, lysis buffers, and hybridization solutions, and is compatible with mass spectrometry, Western blotting, and nucleic acid extraction.
Catalog No. 50-136-8682 Supplier Research Products International Corp Supplier No. U20200500.0
Urea (CO(NH₂)₂) is a water-soluble compound (60.06 g/mol) used in life science labs as a chaotropic agent to denature proteins and nucleic acids. It’s commonly applied in SDS-PAGE, protein solubilization, lysis buffers, and hybridization solutions, and is compatible with mass spectrometry, Western blotting, and nucleic acid extraction.