Learn More
Sigma Aldrich Fine Chemicals Biosciences Urea, 57-13-6, MFCD00008022, 6x25mL
Catalog No. 501657042
Encompass
Linear Formula NH2CONH2, Molecular Weight 60.06, 8M (after reconstitution with 16 mL high purity water), Synonym: Carbamide, Carbonyldiamide, Urea solution; Urea is a chaotropic agent and is used for protein denaturation. It disturbs the hydrogen bonds in the secondary, tertiary and quaternary structure of proteins. It can also disturb hydrogen bonding present in DNA secondary structure.
Catalog No. 50-165-7042 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. U48836X25ML
Linear Formula NH2CONH2, Molecular Weight 60.06, 8M (after reconstitution with 16 mL high purity water), Synonym: Carbamide, Carbonyldiamide, Urea solution; Urea is a chaotropic agent and is used for protein denaturation. It disturbs the hydrogen bonds in the secondary, tertiary and quaternary structure of proteins. It can also disturb hydrogen bonding present in DNA secondary structure.