Learn More
Medchemexpress LLC Urea | 57-13-6 | MFCD00008022 | 99.8% | 60.06 | CH4N2O | 50mg
SDP
Catalog No. 5000267338
Encompass_Preferred
Urea (Standard) is the analytical standard of Urea This product is intended for research and analytical applications Urea is a powerful protein denaturant via both direct and indirect mechanisms[1] A potent emollient and keratolytic agent[2] Used as a diuretic agent Blood urea nitrogen (BUN) has been utilized to evaluate renal function[3] Widely used in fertilizers as a source of nitrogen and is an important raw material for the chemical industry
Catalog No. 50-002-67338 Supplier Medchemexpress LLC Supplier No. HYY0271R50MG
Urea (Standard) is the analytical standard of Urea This product is intended for research and analytical applications Urea is a powerful protein denaturant via both direct and indirect mechanisms[1] A potent emollient and keratolytic agent[2] Used as a diuretic agent Blood urea nitrogen (BUN) has been utilized to evaluate renal function[3] Widely used in fertilizers as a source of nitrogen and is an important raw material for the chemical industry