Learn More
VWR INTERNATIONAL LLC UREA 8M SOLUTN250ML ULTRAPURE
Supplier:  VWR INTERNATIONAL LLC 97062966
Encompass
Commonly used to denature nucleic acids for electrophoresis and to study the secondary and tertiary structure of proteins.
Catalog No. NC0128954