Learn More
Sigma Aldrich Fine Chemicals Biosciences Urea Assay Kit sufficient
Catalog No. 502904333
Encompass
Urea is the major end product of nitrogen metabolism in most animals and is produced in a series of reactions in the liver called the urea cycle. In the urea cycle ammonia is converted to urea which is carried by blood to the kidneys for elimination from the body. High levels of urea in the blood may indicate renal failure. Urea levels may also be elevated in response to treatment with certain drugs such as corticosteroids or in response to decreased kidney filtration due to dehydration or congestive heart failure. Decreased blood urea levels can occur in response to liver disease or malnutrition.
Catalog No. 50-290-4333 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK0061KT
Urea is the major end product of nitrogen metabolism in most animals and is produced in a series of reactions in the liver called the urea cycle. In the urea cycle ammonia is converted to urea which is carried by blood to the kidneys for elimination from the body. High levels of urea in the blood may indicate renal failure. Urea levels may also be elevated in response to treatment with certain drugs such as corticosteroids or in response to decreased kidney filtration due to dehydration or congestive heart failure. Decreased blood urea levels can occur in response to liver disease or malnutrition.