Learn More
Research Products International Corp Urea [Carbamide], UltraPure (ACS Grade), 3 Kilograms

Supplier: Research Products International Corp U202253000.0
Substance Name: Carbamide
CAS Number: 57-13-6
Molecular Formula: CH4N2O
Molecular Weight: 60.1
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.