Learn More
Medchemexpress LLC Urea (carbonyl diamide) | 57-13-6 | MFCD00008022 | 60.06 g/mol | CH4N2O | 1mL
SDP
Catalog No. 5000266954
Encompass_Preferred
Urea is a powerful protein denaturant via both direct and indirect mechanisms[1] A potent emollient and keratolytic agent[2] Used as a diuretic agent Blood urea nitrogen (BUN) has been utilized to evaluate renal function[3] Widely used in fertilizers as a source of nitrogen and is an important raw material for the chemical industry
Catalog No. 50-002-66954 Supplier Medchemexpress LLC Supplier No. HYY027110MM/1ML
Urea is a powerful protein denaturant via both direct and indirect mechanisms[1] A potent emollient and keratolytic agent[2] Used as a diuretic agent Blood urea nitrogen (BUN) has been utilized to evaluate renal function[3] Widely used in fertilizers as a source of nitrogen and is an important raw material for the chemical industry