Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Urea-d3, methyl-d3- | 56683-43-3 | 96.8% | 80.12 g/mol | C2D6N2O | 10 MG
SDP

Catalog No. 5000303130
Encompass_Preferred

1-Methylureal-d6 is the deuterium-labeled analogue of 1-methylurea, provided as an isotopic reference standard for analytical applications such as mass spectrometry and tracer studies. It offers defined isotopic enrichment to support accurate quantitation and method development.

  • Purity 96.8% (HPLC).
  • Isotopic enrichment 96.7%.
  • Molecular formula C2D6N2O.
  • Molecular weight 80.12 g/mol.
  • Appearance white to off-white solid.
  • Storage: powder -20°C (3 years); in solvent -80°C (6 months), -20°C (1 month).
  • Typical pack size 10 mg.

Catalog No. 50-003-03130 Supplier Medchemexpress LLC Supplier No. HYW015971S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Methylureal-d6 is the deuterium-labeled analogue of 1-methylurea, provided as an isotopic reference standard for analytical applications such as mass spectrometry and tracer studies. It offers defined isotopic enrichment to support accurate quantitation and method development.

  • Purity 96.8% (HPLC).
  • Isotopic enrichment 96.7%.
  • Molecular formula C2D6N2O.
  • Molecular weight 80.12 g/mol.
  • Appearance white to off-white solid.
  • Storage: powder -20°C (3 years); in solvent -80°C (6 months), -20°C (1 month).
  • Typical pack size 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.