Learn More
Medchemexpress LLC Urea-d4 | 1433-11-0 | MFCD00044227 | 100.0% | 64.08 g/mol | CD4N2O | 1g
SDP
Catalog No. 5000266956
Encompass_Preferred
Urea-d4 is the deuterium labeled Urea[1] Urea is a powerful protein denaturant via both direct and indirect mechanisms[2] A potent emollient and keratolytic agent[3] Used as a diuretic agent Blood urea nitrogen (BUN) has been utilized to evaluate renal function[4] Widely used in fertilizers as a source of nitrogen and is an important raw material for the chemical industry
Catalog No. 50-002-66956 Supplier Medchemexpress LLC Supplier No. HYY0271S11G
Urea-d4 is the deuterium labeled Urea[1] Urea is a powerful protein denaturant via both direct and indirect mechanisms[2] A potent emollient and keratolytic agent[3] Used as a diuretic agent Blood urea nitrogen (BUN) has been utilized to evaluate renal function[4] Widely used in fertilizers as a source of nitrogen and is an important raw material for the chemical industry