Learn More
MOLECULAR DEPOT LLC Urea For Interference - 2/1 mL
Catalog No. 5000013379
Encompass_Preferred
Urea for Interference Testing consists of a highly concentrated solution of Urea (200 mg/mL) provided with the corresponding diluent (diluent A) This solution can be spiked into specimens to test Urea interference and tolerance in assays
Catalog No. 50-000-13379 Supplier MOLECULAR DEPOT LLC Supplier No. K2010008U
Urea for Interference Testing consists of a highly concentrated solution of Urea (200 mg/mL) provided with the corresponding diluent (diluent A) This solution can be spiked into specimens to test Urea interference and tolerance in assays