Learn More
National Diagnostics UreaGel 29:1 Denaturing Gel System, 1 L
SDP
Catalog No. NC0473760
Encompass
UreaGel 29:1 System allows researchers to easily and safely produce denaturing gels of the popular 29:1 acrylamide:bis-acrylamide formula in concentrations up to 20% monomer. These gels are ideal for analysis of DNA or RNA.
Catalog No. NC0473760 Supplier National Diagnostics Supplier No. EC8291L
UreaGel 29:1 System allows researchers to easily and safely produce denaturing gels of the popular 29:1 acrylamide:bis-acrylamide formula in concentrations up to 20% monomer. These gels are ideal for analysis of DNA or RNA.