Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Urea Hydrogen Peroxide

Catalog No. 289132500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-913-2500G 500 g
28-913-2250G 250 g
28-913-25G 5 g
28-913-2100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 28-913-2500G Supplier Sigma Aldrich Supplier No. 289132500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Urea Hydrogen Peroxide

Specifications

CAS 124-43-6
Melting Point 90°C to 93°C
Molecular Formula CH4N2O -+ H2O2
Linear Formula CO(NH2)2 -+ H2O2
MDL Number MFCD00013119
Quantity 500 g
Synonym Hydrogen peroxide-Urea adduct; Carbamide Per hydrate; Carbamide peroxide; Percarbamide; Urea hydrogen peroxide
Molecular Weight (g/mol) 94.07 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.