Learn More
Medchemexpress LLC Urea, N-1,2-dithiolan-4-yl-N'-[2-oxo-4-(trifluoromethyl)-2H-1-benzopyran-7-yl]- | 2966536-18-3 | 99.7% | 376.37 g·mol⁻¹ | C14H11F3N2O3S2 | 10 MG
SDP
Catalog No. 5000296983
Encompass_Preferred
Fast-TRFS is a selective, superfast fluorogenic probe for thioredoxin reductase (TrxR) that produces fluorescence upon reduction of its disulfide bond, enabling rapid imaging of TrxR activity in live cells. It is supplied as a solid or as a 10 mM solution in DMSO and is suitable for cellular imaging and rapid detection assays.
  • Selective probe for thioredoxin reductase (TrxR).
  • Superfast fluorogenic response upon disulfide reduction.
  • Suitable for live-cell imaging of TrxR activity.
  • Available as 10 mg solid and 10 mM solution in DMSO.
  • High purity (99.7%) and molecular weight 376.37 g·mol⁻¹.
Catalog No. 50-002-96983 Supplier Medchemexpress LLC Supplier No. HYD125210MG
Fast-TRFS is a selective, superfast fluorogenic probe for thioredoxin reductase (TrxR) that produces fluorescence upon reduction of its disulfide bond, enabling rapid imaging of TrxR activity in live cells. It is supplied as a solid or as a 10 mM solution in DMSO and is suitable for cellular imaging and rapid detection assays.
  • Selective probe for thioredoxin reductase (TrxR).
  • Superfast fluorogenic response upon disulfide reduction.
  • Suitable for live-cell imaging of TrxR activity.
  • Available as 10 mg solid and 10 mM solution in DMSO.
  • High purity (99.7%) and molecular weight 376.37 g·mol⁻¹.