Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Urea, N-(2-((4-(diethylamino)butyl)amino)-6-(3,5-dimethoxyphenyl)pyrido(2,3-d)pyrimidin-7-yl)-N'-(1 | 219580-11-7 | MFCD08705327 | 99.9% | 523.7 g/mol | C28H41N7O3 | 200 MG
SDP

Catalog No. 5000247275
Encompass_Preferred

PD173074 is a small-molecule research compound that potently inhibits fibroblast growth factor receptor 1 (FGFR1). It shows an FGFR1 IC50 of ~25 nM, inhibits VEGFR2 with an IC50 of 100-200 nM, and demonstrates approximately 1,000-fold selectivity over PDGFR and c-Src. It is supplied for laboratory research use as an analytical/research standard.

  • Potent FGFR1 inhibitor (IC50 ≈ 25 nM).
  • Also inhibits VEGFR2 (IC50 100-200 nM).
  • Approximately 1,000-fold selectivity for FGFR1 over PDGFR and c-Src.
  • High purity (≈99.9%).
  • Molecular weight 523.7 g/mol.
  • CAS number 219580-11-7.
  • For research use only.

Catalog No. 50-002-47275 Supplier Medchemexpress LLC Supplier No. HY10321200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PD173074 is a small-molecule research compound that potently inhibits fibroblast growth factor receptor 1 (FGFR1). It shows an FGFR1 IC50 of ~25 nM, inhibits VEGFR2 with an IC50 of 100-200 nM, and demonstrates approximately 1,000-fold selectivity over PDGFR and c-Src. It is supplied for laboratory research use as an analytical/research standard.

  • Potent FGFR1 inhibitor (IC50 ≈ 25 nM).
  • Also inhibits VEGFR2 (IC50 100-200 nM).
  • Approximately 1,000-fold selectivity for FGFR1 over PDGFR and c-Src.
  • High purity (≈99.9%).
  • Molecular weight 523.7 g/mol.
  • CAS number 219580-11-7.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.