Learn More
Medchemexpress LLC Urea, N-[2-chloro-7-[(3S,5S)-3,5-dimethyl-4-morpholinyl]pyrazolo[1,5-a]pyrimidin-6-yl]-... | 2682102-10-7 | 99.6% | 469.85 | C19H19ClF3N7O2 | 5 MG
SDP
Catalog No. 5000262908
Encompass_Preferred
MLT-231 is a potent, highly selective allosteric inhibitor of MALT1 intended for research use. It inhibits MALT1 enzymatic activity with IC50 ≈ 9 nM, prevents endogenous BCL10 cleavage (IC50 ≈ 160 nM), and has demonstrated antitumor activity in ABC-DLBCL xenograft models.
  • Potent allosteric inhibition of MALT1 with low-nanomolar activity.
  • Prevents BCL10 cleavage in cellular assays, supporting mechanism studies.
  • Demonstrated antitumor activity in preclinical xenograft models.
  • Available as solid and as DMSO solution for assay flexibility.
  • High chemical purity for reproducible experimental results.
  • Recommended storage conditions support long-term stability.
Catalog No. 50-002-62908 Supplier Medchemexpress LLC Supplier No. HY1319025MG
MLT-231 is a potent, highly selective allosteric inhibitor of MALT1 intended for research use. It inhibits MALT1 enzymatic activity with IC50 ≈ 9 nM, prevents endogenous BCL10 cleavage (IC50 ≈ 160 nM), and has demonstrated antitumor activity in ABC-DLBCL xenograft models.
  • Potent allosteric inhibition of MALT1 with low-nanomolar activity.
  • Prevents BCL10 cleavage in cellular assays, supporting mechanism studies.
  • Demonstrated antitumor activity in preclinical xenograft models.
  • Available as solid and as DMSO solution for assay flexibility.
  • High chemical purity for reproducible experimental results.
  • Recommended storage conditions support long-term stability.