Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Urea, N-[2-(dimethylamino)ethyl]-N'-(7-methylbenzo[1,2-d:3,4-d']bisthiazol-2-yl)- | 1428729-63-8 | 99.5% | 335.45 | C14H17N5OS2 | 50 MG
SDP

Catalog No. 5000241973
Encompass_Preferred

RIG-1 modulator 1 is a small-molecule antiviral research compound described as "Compound of claim 13" with reported activity against influenza virus, hepatitis B virus (HBV), hepatitis C virus (HCV), and human immunodeficiency virus (HIV). It is supplied as a solid (white to light yellow) with molecular formula C14H17N5OS2 and molecular weight 335.45 g/mol for preclinical research use.

  • High purity suitable for research (≈99.5%).
  • Solid form, white to light yellow.
  • Molecular weight 335.45 g/mol.
  • CAS number 1428729-63-8.
  • Reported antiviral activity against influenza, HBV, HCV, and HIV.
  • Available in research sizes, 5 mg-500 mg.

Catalog No. 50-002-41973 Supplier Medchemexpress LLC Supplier No. HY10790250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RIG-1 modulator 1 is a small-molecule antiviral research compound described as "Compound of claim 13" with reported activity against influenza virus, hepatitis B virus (HBV), hepatitis C virus (HCV), and human immunodeficiency virus (HIV). It is supplied as a solid (white to light yellow) with molecular formula C14H17N5OS2 and molecular weight 335.45 g/mol for preclinical research use.

  • High purity suitable for research (≈99.5%).
  • Solid form, white to light yellow.
  • Molecular weight 335.45 g/mol.
  • CAS number 1428729-63-8.
  • Reported antiviral activity against influenza, HBV, HCV, and HIV.
  • Available in research sizes, 5 mg-500 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.