Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Urea, N-[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]-N'-[3-[[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]amino]carbonyl... | 802555-85-7 | MFCD31666371 | 98.6% | 448.52 g/mol | C23H28N8O2 | 100MG
SDP

Catalog No. 5000239104
Encompass_Preferred

p32 Inhibitor M36 is a small-molecule inhibitor that binds directly to the mitochondrial protein p32 and inhibits its association with LyP-1. Supplied as a white to off-white solid for research use only, it is characterized by a molecular formula of C23H28N8O2, a molecular weight of approximately 448.5 g/mol, and high purity to support biochemical and cell-based studies.

  • Inhibits p32 and blocks p32-LyP-1 interaction.
  • Molecular formula C23H28N8O2.
  • Molecular weight 448.5 g/mol.
  • Purity 98.6% (LCMS).
  • Physical form solid, white to off-white powder.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year).
  • Intended for research use only; not for human or clinical use.
  • Available in multiple pack sizes from 5 mg up to 500 mg.

Catalog No. 50-002-39104 Supplier Medchemexpress LLC Supplier No. HY124718100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

p32 Inhibitor M36 is a small-molecule inhibitor that binds directly to the mitochondrial protein p32 and inhibits its association with LyP-1. Supplied as a white to off-white solid for research use only, it is characterized by a molecular formula of C23H28N8O2, a molecular weight of approximately 448.5 g/mol, and high purity to support biochemical and cell-based studies.

  • Inhibits p32 and blocks p32-LyP-1 interaction.
  • Molecular formula C23H28N8O2.
  • Molecular weight 448.5 g/mol.
  • Purity 98.6% (LCMS).
  • Physical form solid, white to off-white powder.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year).
  • Intended for research use only; not for human or clinical use.
  • Available in multiple pack sizes from 5 mg up to 500 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.