Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Urea, N-[(3S,4R)-4-(2,6-difluoro-4-methoxyphenyl)-2-oxo-3-pyrrolidinyl]-N'-phenyl- | 2253947-47-4 | 99.7% | 250 MG
SDP

Catalog No. 5000342808
Encompass_Preferred

This compound is a selective, orally active agonist of formyl peptide receptor 2 (FPR2), exhibiting strong agonistic activity in both human and mouse models. It shows promise in attenuating cardiac remodeling after myocardial infarction, suggesting potential for preventing heart failure. Its mechanism involves inhibiting neutrophil chemotaxis and stimulating macrophage phagocytosis, crucial for resolving inflammation. Suitable for in vitro and in vivo research, it demonstrates good oral bioavailability in animal models.

  • Selective agonist of formyl peptide receptor 2 (FPR2).
  • Highly selective for FPR2 (human EC50 0.41 nM, mouse EC50 3.4 nM).
  • Inhibits neutrophil chemotaxis; promotes macrophage phagocytosis.
  • Attenuates cardiac remodeling after myocardial infarction.
  • Orally active with 24% bioavailability in mice.
  • Purity: 99.74%.
  • Molecular formula: C18H17F2N3O3.
  • Molecular weight: 361.34.

Catalog No. 50-003-42808 Supplier Medchemexpress LLC Supplier No. 250MGHY131180
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This compound is a selective, orally active agonist of formyl peptide receptor 2 (FPR2), exhibiting strong agonistic activity in both human and mouse models. It shows promise in attenuating cardiac remodeling after myocardial infarction, suggesting potential for preventing heart failure. Its mechanism involves inhibiting neutrophil chemotaxis and stimulating macrophage phagocytosis, crucial for resolving inflammation. Suitable for in vitro and in vivo research, it demonstrates good oral bioavailability in animal models.

  • Selective agonist of formyl peptide receptor 2 (FPR2).
  • Highly selective for FPR2 (human EC50 0.41 nM, mouse EC50 3.4 nM).
  • Inhibits neutrophil chemotaxis; promotes macrophage phagocytosis.
  • Attenuates cardiac remodeling after myocardial infarction.
  • Orally active with 24% bioavailability in mice.
  • Purity: 99.74%.
  • Molecular formula: C18H17F2N3O3.
  • Molecular weight: 361.34.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.